Tisk…

Anti-Ly-6G Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB13552-100 ORB13552-200
BIRBORB13552-100EA 0 CZK
BIRBORB13552-100 BIRBORB13552-200
Anti-Ly-6G Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Ly-6G (Ly-6G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR