Tisk…
Anti-Glu-Glu Tag Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB13348-100 ORB13348-200
BIRBORB13348-100EA 0 CZK
BIRBORB13348-100 BIRBORB13348-200
Anti-Glu-Glu Tag Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Glu-Glu Tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat