Tisk…

Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
ORB12018-100 ORB12018-50
BIRBORB12018-100EA 0 CZK
BIRBORB12018-100 BIRBORB12018-50
Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR