Tisk…

Anti-ROCK1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB11336-100 ORB11336-200
BIRBORB11336-100EA 0 CZK
BIRBORB11336-100 BIRBORB11336-200
Anti-ROCK1 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: ROCK1 (Rho-associated, coiled-coil containing protein kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR