Tisk…
Anti-B2M Mouse Monoclonal Antibody
Dodavatel: Biorbyt
ORB11066-100 ORB11066-200
BIRBORB11066-100EA 0 CZK
BIRBORB11066-100 BIRBORB11066-200
Anti-B2M Mouse Monoclonal Antibody
Protilátky
  • ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human