Tisk…

Anti-CXCR3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB10696-100 ORB10696-200
BIRBORB10696-100EA 0 CZK
BIRBORB10696-100 BIRBORB10696-200
Anti-CXCR3 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CXCR3 (chemokine (C-X-C motif) receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR