Tisk…

Anti-Dopamine D1 receptor Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB10514-100 ORB10514-20 ORB10514-200 ORB10514-5 ORB10514-50
BIRBORB10514-100EA 0 CZK
BIRBORB10514-100 BIRBORB10514-20 BIRBORB10514-200 BIRBORB10514-5 BIRBORB10514-50
Anti-Dopamine D1 receptor Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Dopamine D1 receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR