Tisk…

Anti-COX II Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB10450-200 ORB10450-100
BIRBORB10450-200EA 0 CZK
BIRBORB10450-200 BIRBORB10450-100
Anti-COX II Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: COX II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR