Tisk…
Anti-BOB1 Rabbit polyclonal antibody
Dodavatel: Biorbyt
ORB10199-200 ORB10199-100
BIRBORB10199-200EA 0 CZK
BIRBORB10199-200 BIRBORB10199-100
Anti-BOB1 Rabbit polyclonal antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: BOB1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat