Tisk…
Anti-ACVR2A Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB10137-100 ORB10137-200
BIRBORB10137-100EA 0 CZK
BIRBORB10137-100 BIRBORB10137-200
Anti-ACVR2A Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ACVR2A (activin A receptor, type IIA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat