Tisk…

Anti-2,4-Dichlorophenoxyacetic acid Mouse Monoclonal Antibody

Dodavatel: Biorbyt
ORB10005-100 ORB10005-200
BIRBORB10005-100EA 0 CZK
BIRBORB10005-100 BIRBORB10005-200
Anti-2,4-Dichlorophenoxyacetic acid Mouse Monoclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: 2,4-Dichlorophenoxyacetic acid
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR