Tisk…

Anti-TCEB2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB7076-100 ORB7076-200
BIRBORB7076-100EA 0 CZK
BIRBORB7076-100 BIRBORB7076-200
Anti-TCEB2 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: TCEB2 (transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR