Tisk…
Anti-AFP Mouse Monoclonal Antibody [clone: A4]
Dodavatel: Biorbyt
ORB4246-100 ORB4246-200
BIRBORB4246-100EA 0 CZK
BIRBORB4246-100 BIRBORB4246-200
Anti-AFP Mouse Monoclonal Antibody [clone: A4]
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Flow Cytometry: 1:20-100; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: monoclonal
Clone: A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human