Tisk…

Anti-C6orf58 Rabbit polyclonal antibody

Dodavatel: Biorbyt
ORB2969-100 ORB2969-200
BIRBORB2969-100EA 0 CZK
BIRBORB2969-100 BIRBORB2969-200
Anti-C6orf58 Rabbit polyclonal antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: C6orf58 (chromosome 6 open reading frame 58)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR