Tisk…

Anti-BKCA alpha Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB2919-100 ORB2919-200
BIRBORB2919-100EA 0 CZK
BIRBORB2919-100 BIRBORB2919-200
Anti-BKCA alpha Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: BKCA alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR