Tisk…

Anti-CTBP1-AS2 Rabbit polyclonal antibody

Dodavatel: Biorbyt
ORB2845-100 ORB2845-200
BIRBORB2845-100EA 0 CZK
BIRBORB2845-100 BIRBORB2845-200
Anti-CTBP1-AS2 Rabbit polyclonal antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CTBP1-AS2 (CTBP1 antisense RNA 2 (head to head))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR