Tisk…
Anti-HHLA2 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB2138-100 ORB2138-200
BIRBORB2138-100EA 0 CZK
BIRBORB2138-100 BIRBORB2138-200
Anti-HHLA2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: HHLA2 (HERV-H LTR-associating 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human