Tisk…

Anti-DHFRL1 Rabbit polyclonal antibody

Dodavatel: Biorbyt
ORB2052-100 ORB2052-200
BIRBORB2052-100EA 0 CZK
BIRBORB2052-100 BIRBORB2052-200
Anti-DHFRL1 Rabbit polyclonal antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: DHFRL1 (dihydrofolate reductase-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR