Tisk…

Anti-OST beta Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB1964-100 ORB1964-200
BIRBORB1964-100EA 0 CZK
BIRBORB1964-100 BIRBORB1964-200
Anti-OST beta Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: OST beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR