Tisk…

Anti-HPRT1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB1943-100 ORB1943-200
BIRBORB1943-100EA 0 CZK
BIRBORB1943-100 BIRBORB1943-200
Anti-HPRT1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: HPRT1 (hypoxanthine phosphoribosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR