Tisk…

Anti-RNF32 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB1473-100 ORB1473-200
BIRBORB1473-100EA 0 CZK
BIRBORB1473-100 BIRBORB1473-200
Anti-RNF32 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RNF32 (ring finger protein 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR