Tisk…
Anti-ART5 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB1309-100 ORB1309-200
BIRBORB1309-100EA 0 CZK
BIRBORB1309-100 BIRBORB1309-200
Anti-ART5 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: ART5 (ADP-ribosyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat