Tisk…

Anti-AK9 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB1271-100 ORB1271-200
BIRBORB1271-100EA 0 CZK
BIRBORB1271-100 BIRBORB1271-200
Anti-AK9 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: AK9 (adenylate kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR