Tisk…

Anti-KLHDC9 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB1248-100 ORB1248-200
BIRBORB1248-100EA 0 CZK
BIRBORB1248-100 BIRBORB1248-200
Anti-KLHDC9 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: KLHDC9 (kelch domain containing 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR