Tisk…

Anti-AHCTF1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB1089-100 ORB1089-200
BIRBORB1089-100EA 0 CZK
BIRBORB1089-100 BIRBORB1089-200
Anti-AHCTF1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: AHCTF1 (AT hook containing transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR