Tisk…

Anti-LRRC32 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: GARP5]

Dodavatel: Tonbo Biosciences
50-9882-T025 50-9882-T100
50-9882-T025EA 3310 CZK
50-9882-T025 50-9882-T100
Anti-LRRC32 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: GARP5]
Protilátky
The GARP5 antibody reacts with human GARP, also known as LRRC32 or Garpin, an 80 kDa transmembrane protein which acts as a receptor for the latent form of TGF-beta 1 (pro-TGF-beta), preventing its secretion. Specifically, GARP is reported to associate with the pro-domain of TGF-beta 1, known as latency-associated peptide (LAP), which must be cleaved to release the biologically active cytokine. The putative role of GARP in sequestering the TGF-beta is important for regulating its activity, as TGF-beta signaling is involved in development and function of Th17, Treg and many other immune cell types. GARP expression has been shown on platelets and at high levels on Foxp3+ Treg cells, where it is proposed to be a phenotypic identifier for activated human Treg cells.

The GARP5 antibody may be used for analysis of human GARP by flow cytometry, immunoprecipitation and Western blotting. Please choose the appropriate format for each application.

Type: Primary
Antigen: LRRC32 (leucine rich repeat containing 32)
Clonality: monoclonal
Clone: GARP5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR