Tisk…

Anti-TNFRSF18 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DTA-1]

Dodavatel: Tonbo Biosciences
50-5874-U100 50-5874-U025
50-5874-U100EA 5350 CZK
50-5874-U100 50-5874-U025
Anti-TNFRSF18 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DTA-1]
Protilátky
The DTA-1 antibody reacts with mouse CD357, also known as GITR or AITR (in humans), a 66-70 kDa member of the Tumor Necrosis Factor superfamily (TNFRSF18). GITR is primarily found on T cells, and its function may vary depending on the T cell type where it is expressed. GITR is upregulated on activated T cells where it provides co-stimulation, yet GITR may promote the inhibition of CD4+ CD25+ Treg cells, where it is expressed at high levels. GITR ligand (GITRL) is found on B cells, macrophages, dendritic and endothelial cells, and is implicated in regulating both innate and adaptive immune responses.

The DTA-1 antibody may be used for analysis of GITR expression on T cells, and is also commonly used in vitro as an agonistic antibody to induce GITR signaling in various assays.

Type: Primary
Antigen: TNFRSF18 (tumor necrosis factor receptor superfamily, member 18)
Clonality: monoclonal
Clone: DTA-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR