Tisk…

Anti-CD45.1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: A20]

Dodavatel: Tonbo Biosciences
35-0453-U500 35-0453-U025 35-0453-U100
35-0453-U500EA 3220 CZK
35-0453-U500 35-0453-U025 35-0453-U100
Anti-CD45.1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: A20]
Protilátky
The A20 antibody reacts with mouse CD45.1, also known as Ly5.1, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.

The A20 antibody is typically used as a leukocyte marker in Ly5.1 mouse strains: SJL/J, DA, STS/A and RIII. The antibody has been demonstrated to specific for CD45.1 and is not cross-reactive with CD45.2 bearing cells.

Type: Primary
Antigen: CD45.1 (CD45.1)
Clonality: monoclonal
Clone: A20
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR