Tisk…
Anti-ITGAX Armenian Hamster Monoclonal Antibody (Biotin) [clone: N418]
Dodavatel: Tonbo Biosciences
30-0114-U500 30-0114-U100 30-0114-U025
30-0114-U500EA 4870 CZK
30-0114-U500 30-0114-U100 30-0114-U025
Anti-ITGAX Armenian Hamster Monoclonal Antibody (Biotin) [clone: N418]
Protilátky
The N418 antibody reacts with mouse CD11c, also known as integrin alpha X. This 150 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18)More Product Information