Tisk…

Anti-MHC II (I-A/I-E) Rat Monoclonal Antibody (APC (Allophycocyanin) [clone: M5/115.15.2]

Dodavatel: Tonbo Biosciences
20-5321-U100 20-5321-U025
20-5321-U100EA 3820 CZK
20-5321-U100 20-5321-U025
Anti-MHC II (I-A/I-E) Rat Monoclonal Antibody (APC (Allophycocyanin) [clone: M5/115.15.2]
Protilátky
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.

The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.

Type: Primary
Antigen: MHC II (I-A/I-E) (MHC II (I-A/I-E))
Clonality: monoclonal
Clone: M5/115.15.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR