Tisk…
Anti-USE1 Rabbit Polyclonal Antibody
Dodavatel: Aviva Systems Biology
ARP37352_P050 ARP37352_T100
AVIVARP37352_P050EA 11290 CZK
AVIVARP37352_P050 AVIVARP37352_T100
Anti-USE1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: USE1 (unconventional SNARE in the ER 1 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat