Tisk…
Anti-SERTAD1 Rabbit Polyclonal Antibody
Dodavatel: Aviva Systems Biology
ARP34309_P050 ARP34309_T100
AVIVARP34309_P050EA 11290 CZK
AVIVARP34309_P050 AVIVARP34309_T100
Anti-SERTAD1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SERTAD1 (SERTA domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat