Tisk…
Anti-PARP11 Rabbit Polyclonal Antibody
Dodavatel: Aviva Systems Biology
ARP33768_P050 ARP33768_T100
AVIVARP33768_P050EA 11290 CZK
AVIVARP33768_P050 AVIVARP33768_T100
Anti-PARP11 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PARP11 (poly (ADP-ribose) polymerase family, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat