Tisk…
Capillary columns, DB-1701
Dodavatel: Agilent
121-0722 121-0722LTM 122-0713LTM 122-0732LTM 122-0733LTM 123-0712LTM 125-0712LTM
HEWL121-0722EA 21950 CZK
HEWL121-0722 HEWL121-0722LTM HEWL122-0713LTM HEWL122-0732LTM HEWL122-0733LTM HEWL123-0712LTM HEWL125-0712LTM
Capillary columns, DB-1701
Kolony chromatografické
These columns contain (14%-Cyanopropyl-phenyl)-methylpolysiloxane.
  • Low/mid-polarity
  • Bonded and cross-linked
  • Solvent rinsable