Tisk…
Capillary columns, DB-5ms Ultra Inert
Dodavatel: Agilent
121-5522UI 121-5522UILTM 121-5523UILTM 122-5512UILTM G3900-63005 165-6626 221-5522UILTM
HEWL121-5522UIEA 25900 CZK
HEWL121-5522UI HEWL121-5522UILTM HEWL121-5523UILTM HEWL122-5512UILTM HEWLG3900-63005 HEWL165-6626 HEWL221-5522UILTM
Capillary columns, DB-5ms Ultra Inert
Kolony chromatografické
Recommended columns for semivolatiles, halogenated compounds, pesticides, herbicides, drugs of abuse, amines, unknown sample screening.
5% Phenyl 95% dimethyl arylene siloxane.