Tisk…
Sampling scoops, LaboPlast®
Dodavatel: BURKLE
5378-0001 5378-0002 5378-0003 5378-0004 5378-0005 5378-0006 5378-0007 5378-0008 5378-0013 5378-0014 5378-0015 5378-0016 5378-0019 5378-0020
231-0503EA 555 CZK
231-0503 231-0504 231-0505 231-0506 231-0507 231-0508 231-0509 231-0510 BURK5378-0013 BURK5378-0014 BURK5378-0015 BURK5378-0016 BURK5378-0019 331-0365
Sampling scoops, LaboPlast®
Lopatky
PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediately after the sampling procedure to secure a clean and contamination-free sample.
  • Individually packed
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.