Tisk…
Examination gloves, latex, Unigloves® Safetec
Dodavatel: UNIGLOVES AERTZE UND KLINIKBED
1302 1303 1304 1305
112-1779EA 932 CZK
112-1779 112-1780 112-1781 112-1782
Examination gloves, latex, Unigloves® Safetec
Rukavice
These ambidextrous gloves are suitable for handling food.
  • Non sterile, powder-free
  • Beaded cuffs
  • Micro-roughened palm
Length: 240 mm
Thickness: 0,11 - 0,15 mm (cuff); 0,15 - 0,18 mm (palm); 0,18 - 0,22 mm (finger)
Colour: White
EN 455, AQL 1,5
Certifikace: EN 455, AQL 1,5