Tisk…

Oxid vápenatý, powder, BAKER ANALYZED®, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

1410-05EA 10260 CZK
1410-05 1410-07 1410-01
Oxid vápenatý, powder, BAKER ANALYZED®, J.T.Baker®
Oxid vápenatý

Powder. Lot analysis on label.

Vzorec: CaO
MW: 56.08
Bod varu: 2850
Bod tání: 2580
Hustota: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS číslo: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use = 1.0 %
Insoluble in CH₃COOH and NH₄OH Precipitate ≤ 1.0 %
Loss on Ignition ≤ 5.0 %
Chloride (Cl) ≤ 0.005 %
Nitrate (NO₃) ≤ 0.05 %
Sulfate (SO₄) ≤ 0.1 %
Heavy Metals (as Pb) ≤ 0.005 %
Iron (Fe) ≤ 0.1 %
Assay, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR