Tisk…
Dip slides
Dodavatel: THERMO FISHER DIAGNOSTICS
DS0147A DS0155A DS0166A DS0167A DS0168A DS0169A DS0170A GFD04A GFD06A GFD07A
OXOIDS0147AEA 1270 CZK
OXOIDS0147A OXOIDS0155A OXOIDS0166A OXOIDS0167A OXOIDS0168A OXOIDS0169A OXOIDS0170A OXOIGFD04A OXOIGFD06A OXOIGFD07A
Dip slides
Média mikrobiologická Připravená mikrobiologická média
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple