Tisk…
Capillary columns, HP-PLOT MoleSieve
Dodavatel: Agilent
19095P-MS5 19095P-MS9 19091P-MS8LTM 19095P-MS0LTM
HEWL19095P-MS5EA 21380 CZK
HEWL19095P-MS5 HEWL19095P-MS9 HEWL19091P-MS8LTM HEWL19095P-MS0LTM
Capillary columns, HP-PLOT MoleSieve
Kolony chromatografické
For applications requiring resolution of argon and oxygen without cryogenic cooling, select columns with thick film. The thin film configurations are ideal for many applications, including routine air monitoring and analysis. The thin film column resolves argon and oxygen at subambient temperatures.