Tisk…
Glove liners, Puri ESD Top Fit
Dodavatel: PURITECH CO LTD
TFG600ESD/S TFG600ESD/XL TFG600ESD/M TFG600ESD/L TFG600ESD/XXL
112-6220EA 2390 CZK
112-6220 112-6221 112-6219 112-6218 112-6222
Glove liners, Puri ESD Top Fit
Rukavice
These thin glove liners made of 100% soft nylon fabric with carbon and PU coating on the fingertips are suitable for handling electronics.
  • Hand-specific for great fit
  • Excellent dexterity
  • Anti-static, prevent damaging static discharge
  • Protect humans and products from static electricity
Ideal for assembling LSI , VLSI, semiconductors as well as computers and precision instruments.
Thickness of coating: 0,1 mm.