Tisk…
In-line filters, SSI™
Dodavatel: Merck
59262-U 59264 59265
SIAL59262-UEA 5920 CZK
SIAL59262-U SIAL59264 SIAL59265
In-line filters, SSI™
Filtry Filtry In-Line
The high pressure in-line filter should be place between the pump and injector to provide final filtration for the mobile phase.
Maximum operating pressure: 1034,21 bar