Tisk…
GC capillary column connectors, Finger Tite
Dodavatel: Thermo Fisher Scientific
290SA131 290SA132 290SA133 290ST130 290S1132 290S1131
555-9149EA 13130 CZK
555-9149 555-9148 555-9147 555-9182 555-9151 555-9152
GC capillary column connectors, Finger Tite
Fitinky a konektory
Metal-to-metal seal eliminates contamination and gives lower air/water background noise. No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures.
  • Leak free connections
  • No tools required