Tisk…
Glove liners, Puri Liner Fit
Dodavatel: PURITECH
IG510-S IG510-L IG510-M IG510-XL IG500-L IG510-XL10CM IG510-M10CM IG510XXL IG500-M IG500-XL IG500-S IG510SINDPACKE IG510MINDPACKE IG510LINDPACKE IG510XLINDPACK IG510XXLINDPAC IG510-L10CM IG510-XXL10CM
112-6584EA 4260 CZK
112-6584 112-6580 112-6582 112-6585 112-6576 112-6586 112-6583 PURTIG510XXL 112-6577 112-6579 112-6578 PURTIG510SINDPACKE PURTIG510MINDPACKE PURTIG510LINDPACKE PURTIG510XLINDPACK PURTIG510XXLINDPAC 112-6581 112-6587
Glove liners, Puri Liner Fit
Rukavice
These glove liners made of 100% soft nylon fabric are ideal for handling electronics. When used as inner gloves, they are also suitable for cleanroom environments.
  • Hand-specific for great fit
  • Excellent dexterity
  • High absorbency
  • Colour-coded cuff indicates size
  • Available with extra long cuff for additional protection
Weight: 16 to 19 g/m²
Weight: 16 - 19 g/m²
Colour: White