Tisk…
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
Dodavatel: PanReac AppliChem
Synonyms: 2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl
A1001.0010 A1001.0100
A1001.0010EA 2090 CZK
A1001.0010 A1001.0100
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
DAPI (4',6-diamidine-2-phenylindole dihydrochloride) is an excellent dye for staining DNA.
DAPI is used as a reagent to detect mycoplasma or virus DNA in cell culture.
Vzorec: C₁₆H₁₅N₅·2HCl
MW: 350.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012681
CAS číslo: 28718-90-3