Tisk…
Precision balances, FX-i, FX-iWP, FZ-i and FZ-iWP series
Dodavatel: AnD Instruments Ltd (A&D)
FZ-1200I-WP-EC FX-2000I-WP FX-200I FX-200I-WP FX-5000I FX-500I FZ-1200I-EC FZ-120I-EC FZ-120I-WP-EC FZ-2000I-EC FZ-2000I-WP-EC FZ-200I-EC FZ-200I-WP-EC FZ-3000I-EC FZ-3000I-WP-EC FZ-300I-EC FZ-300I-WP-EC FX-2000I FZ-500I
611-3038EA 0 CZK
611-3038 611-3033 611-2864 611-3035 611-2870 611-2866 611-2875 611-2871 611-3039 611-2876 611-3040 611-2872 611-3041 611-2877 611-3042 611-2873 611-3043 611-2868 611-2874
Precision balances, FX-i, FX-iWP, FZ-i and FZ-iWP series
Váhy
These compact and lightweight precision balances from A&D are designed to offer a cost effective solution, whilst still delivering great performance and quality.
  • Multiple weighing units including one custom unit
  • Vacuum fluorescence display
  • Choice of internal or external calibration
  • RS232 bi-directional interface
  • GLP conformity with output of ID and calibration data
Units feature all metal construction, stainless steel pan and quick stabilisation times (approx. 1 s). Applications include counting, percentage weighing, animal weighing and below balance weighing. All models feature auto-off function, overload protection, standard comparator function (HI/OK/LO annunciator) and counting function with Automatic Counting Accuracy Improvement. The balances also feature built-in statistical calculations function allowing statistical data to be output or printed to a standard printer.
FZ-i and FZ-iWP balances feature internal calibration. FZ-iWP and FX-iWP models are IP 65 compliant: Water and dust-proof.
Informace pro objednávání: Please contact your local Avantor sales office for details of accessories including USB interface, rechargeable Ni-MH battery, calibration weights, animal weighing pan, Ethernet interface with WinCT-Plus software and waterproof RS232 cable for FX/Z-i-WP balances.
Informace o dodávce: Supplied with AC adapter. All 1 mg models are supplied with a draught shield.
Upozornení: Please note these products are not available in Republic of Ireland or Northern Ireland.