Tisk…
Moulded pathology media vials, WHEATON®
Dodavatel: DWK Life Sciences
128139S 128044F
LSUK128139SEA 2350 CZK
LSUK128139S LSUK128044F
Moulded pathology media vials, WHEATON®
Vialky
Manufactured from neutral Type 1B glass. These vials are suitable for all pharmaceutical, media, diagnostic and sample collection applications.
  • Thick walled moulded glass, chemically inert and with excellent thermal properties
  • Aluminium caps fitted with non-inhibitory rubber liners for storage of cultures and allows cell growth
  • Self sealing PP caps provide leak proof seal