Tisk…
Safety shoes, lace-up, Compositelite™ ULTRA Vistula Trainer, FC67
Dodavatel: Portwest
FC67BKR42 FC67BKR41 FC67BKR40 FC67BKR39 FC67BKR38 FC67BKR45 FC67BKR43 FC67BKR44 FC67BKR47 FC67BKR46
PRTWFC67BKR42EA 2470 CZK
PRTWFC67BKR42 PRTWFC67BKR41 PRTWFC67BKR40 PRTWFC67BKR39 PRTWFC67BKR38 PRTWFC67BKR45 PRTWFC67BKR43 PRTWFC67BKR44 PRTWFC67BKR47 PRTWFC67BKR46
Safety shoes, lace-up, Compositelite™ ULTRA Vistula Trainer, FC67
Obuv
Metal-free, anti-static safety shoes made from micro PU upper plus a durable, dual-density phylon/rubber outer sole, resistant to oil and heat (300 °C).
  • Shock-absorbent heel
  • Composite toecap (200 J)
  • Puncture resistant, composite midsole
Suitable for light industrial use.
Colour: Black/blue
Certifikace: EN 20345:2011 + A1:2007; S1P, SRC.