Tisk…
Capillary columns for specific EPA methods, Trace™
Dodavatel: Thermo Fisher Scientific
26RF223P 26RV495P 26RX142P
555-9061EA 27750 CZK
555-9061 555-9059 555-9058
Capillary columns for specific EPA methods, Trace™
Kolony chromatografické
These columns are designed to perform specific EPA methodologies. These low-bleed and temperature-stable columns have been designed, manufactured and tested to provide consistent, reliable results from run-to-run and batch-to-batch.
  • Low bleed, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
Applications: Volatile organic compounds (VOCs), pesticides, brominated flame retardants, explosives.