Tisk…
HPLC columns, Discovery C18, Supelco®
Dodavatel: Merck
504971-30 504955-30 504955-40 504971-40
504971-30EA 26000 CZK
504971-30 504955-30 504955-40 504971-40
HPLC columns, Discovery C18, Supelco®
Kolony chromatografické
Discovery® C18 columns are designed for the separation of peptides and small proteins with exceptional peak shape for basic and acidic analytes.
  • Excellent reproducibility
  • Suitable for LC/MS application
  • Lower hydrophobicity, providing faster analysis
Stable pH: 2,0 to 8,0
Informace o dodávce: Validation packs includes three columns, each from a different lot of bonded phase.
Supelguard™ guard cartridge kit includes one cartridge, a stand-alone holder, a piece of tubing, two nuts and ferrules.